Order Entry
Belgium
ContactUsLinkComponent
Copper standard solution, 5000 mg/L Cu in 1 M nitric acid (from Cu(NO₃)₂.3H₂O) standard for AAS
Copper standard solution, 5000 mg/L Cu in 1 M nitric acid (from Cu(NO₃)₂.3H₂O) standard for AAS
Catalog # KRAF03611.2000
Supplier:  Bernd Kraft
CAS Number:  
Copper standard solution, 5000 mg/L Cu in 1 M nitric acid (from Cu(NO₃)₂.3H₂O) standard for AAS
Catalog # KRAF03611.2000
Supplier:  Bernd Kraft
CAS Number:  

Specifications

  • Concentration:
    5000 mg/L
  • Matrix:
    1 M HNO₃
  • Pack type:
    Plastic bottle
  • Pk:
    500 mL
  • Element:
    Copper
  • Symbol:
    Cu

Specifications

About this item

Certifications: Traceable to NIST