Order Entry
Belgium
ContactUsLinkComponent
 
Potassium standard solution, 5000 mg/L K in 1 M nitric acid (from KNO₃) standard for AAS
  KRAF03342.2000
 :  
Potassium standard solution, 5000 mg/L K in 1 M nitric acid (from KNO₃) standard for AAS
  KRAF03342.2000
 :  

 

  • Concentration:
    5000 mg/L
  • Matrix:
    1 M HNO₃
  • Pack type:
    Plastic bottle
  • Pk:
    500 mL
  • Element:
    Potassium
  • Symbol:
    K

 

 

 : Traceable to NIST