Potassium standard solution, 10,000 mg/L K in 2 M nitric acid (from KNO₃) standard for AAS
Catalog # KRAF19246.0000
Supplier: Bernd Kraft
CAS Number:
Specifications
- Concentration:10000mg/L
- Matrix:2 M HNO₃
- Pack type:Plastic bottle
- Pk:100 mL
- Element:Potassium
- Symbol:K