Calcium standard solution, 5000 mg/L Ca in 1 M nitric acid (from Ca(NO₃)₂.4H₂O) standard for AAS
Catalog # KRAF03630.2000
Supplier: Bernd Kraft
CAS Number:
Specifications
- Concentration:5000 mg/L
- Matrix:1 M HNO₃
- Pack type:Plastic bottle
- Pk:500 mL
- Element:Calcium
- Symbol:Ca
Specifications
About this item
Certifications: Traceable to NIST