Order Entry
Belgium
ContactUsLinkComponent
Calcium standard solution, 10000 mg/L Ca in 2 M nitric acid (from CaCO3) standard for AAS
Calcium standard solution, 10000 mg/L Ca in 2 M nitric acid (from CaCO3) standard for AAS
Catalog # KRAF20403.0000
Supplier:  Bernd Kraft
CAS Number:  
Calcium standard solution, 10000 mg/L Ca in 2 M nitric acid (from CaCO3) standard for AAS
Catalog # KRAF20403.0000
Supplier:  Bernd Kraft
CAS Number:  

Specifications

  • Concentration:
    10000 mg/L
  • Matrix:
    2 M HNO₃
  • Pack type:
    Plastic bottle
  • Pk:
    100 mL
  • Element:
    Calcium
  • Symbol:
    Ca

Specifications

About this item