Order Entry
Belgium
ContactUsLinkComponent
Pesticide reference standard, Clofenotane 100 µg/ml in n-nonane, CIL
Pesticide reference standard, Clofenotane 100 µg/ml in n-nonane, CIL
Catalog # PROOCILULM61351.2
CAS Number:  
Pesticide reference standard, Clofenotane 100 µg/ml in n-nonane, CIL
Catalog # PROOCILULM61351.2
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    1,2 mL
  • CAS No.:
    50-29-3
  • Description:
    Clofenotane 100 µg/ml in n-nonane
  • Formula:
    C₁₄H₉Cl₅
  • Description:
    Clofenotane

Specifications

About this item