Order Entry
Belgium
ContactUsLinkComponent
 
Mercury standard solution, 5000 mg/L Hg in 1 M nitric acid (from Hg(NO₃)₂) standard for AAS
Catalog # KRAF03445.2000
Supplier:  Bernd Kraft
CAS Number:  
Mercury standard solution, 5000 mg/L Hg in 1 M nitric acid (from Hg(NO₃)₂) standard for AAS
Catalog # KRAF03445.2000
Supplier:  Bernd Kraft
CAS Number:  

Specifications

  • Concentration:
    5000 mg/L
  • Matrix:
    1 M HNO₃
  • Pack type:
    Plastic bottle
  • Pk:
    500 mL
  • Element:
    Mercury
  • Symbol:
    Hg

Specifications

About this item

Certifications: Traceable to NIST