Sodium standard solution, 5000 mg/L Na in 1 M nitric acid (from NaNO₃) standard for AAS
Catalog # KRAF03627.2000
Supplier: Bernd Kraft
CAS Number:
Specifications
- Concentration:5000 mg/L
- Matrix:1 M HNO₃
- Pack type:Glass bottle
- Pk:500 mL
- Element:Sodium
- Symbol:Na
Specifications
About this item
Certifications: Traceable to NIST