Sodium standard solution, 10000 mg/L Na in 2 M nitric acid (from NaNO₃) standard for AAS
Catalog # KRAF12484.0000
Supplier: Bernd Kraft
CAS Number:
Specifications
- Concentration:10000 mg/L
- Matrix:2 M HNO₃
- Pack type:Plastic bottle
- Pk:100 mL
- Element:Sodium
- Symbol:Na