
Sulphuric acid 9 mol/l (18 N)
Catalog # FFCHFF60711000
Supplier: FF CHEMICALS
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:1 L
- Seq:5037
Specifications
About this item
:
:98.08 g/mol
Storage Temperature:Ambient
EINECS:231-639-5
UN:2796
ADR:8,II
:GHS05
CAS Number:7664-93-9
Formula:H₂SO₄
MDL Number:MFCD00064589