Order Entry
Belgium
ContactUsLinkComponent
 
Palladium standard solution, 1,000 mg/L Pd in 10% hydrochloric acid standard for ICP
Catalog # REGEPPD2A8
Supplier:  REAGECON
CAS Number:  
Palladium standard solution, 1,000 mg/L Pd in 10% hydrochloric acid standard for ICP
Catalog # REGEPPD2A8
Supplier:  REAGECON
CAS Number:  

Specifications

  • Concentration:
    1000 mg/L
  • Matrix:
    10% HCl
  • Pack type:
    Glass bottle
  • Pk:
    100 mL
  • Element:
    Palladium
  • Symbol:
    Pd

Specifications

About this item