ContactUsLinkComponent
Fluoride standard solution, Titrisol®, Supelco®
Fluoride standard solution, Titrisol®, Supelco®
  1.09869.0001
 :  Merck
 :  
Fluoride standard solution, Titrisol®, Supelco®
  1.09869.0001
 :  Merck
 :  
  • Concentration:
    1000 mg/l
  • Pack type:
    Plastic ampoule
  • Pk:
    1 Ampoul
  • Description:
    Fluoride standard solution
  • Contents:
    KF in H₂O