Dimethyl sulphoxide, Isocratic Grade for HPLC, for spectroscopy
KRAF19933.4700
:
- Pack type:Glass bottle
- Pk:2.500 mL
- Seq:2895
:
:78.14 g/mol
:13,03285
:189 °C (1013 hPa)
:18.5 °C
:1.101 g/cm³ (20 °C)
:87 °C
:Ambient
:200-664-3
:67-68-5
:(CH₃)₂SO
:MFCD00002089