Orders LinkContactUsLinkComponent
Curd butyrometer, without stopper
Curd butyrometer, without stopper
Butyrometer for curd, 0 - 20 % : 0,2
Catalog # 614-0134
Supplier:   GERBER FUNKE
Curd butyrometer, without stopper
Catalog # 614-0134
Supplier:  GERBER FUNKE
Curd butyrometer, without stopper
Curd butyrometer, without stopper
Butyrometer for curd, 0 - 20 % : 0,2
Price
187,00 €
Log in for availability
1 * 10 ST
Product Details & Documents
Weighing method.
Specifications
  • Description:
    Butyrometer for curd, 0 - 20 % : 0,2
Accessories
Product Information
  • Catalog #
    Availability
    Price
  • Cheese beaker, perforated, short design, for butyrometers acc. to Van Gulik´s method
    Cheese beaker, perforated, short design, for butyrometers acc. to Van Gulik´s method
    614-0141
    Log in for availability
    1 * 1 ST - 9,80 €
Product Information
  • Catalog #
    Availability
    Price
  • Stopper, rubber, with hole, for all weighing method butyrometers
    Stopper, rubber, with hole, for all weighing method butyrometers
    614-2030
    Log in for availability
    1 * 100 ST - 72,30 €
Product Information
  • Catalog #
    Availability
    Price
  • Cheese beaker, perforated, for butyrometers acc. to Van Gulik´s method
    Cheese beaker, perforated, for butyrometers acc. to Van Gulik´s method
    614-2033
    Log in for availability
    1 * 1 ST - 4,20 €
Product Information
  • Catalog #
    Availability
    Price
  • Stopper, rubber, for sealing the bulbs of all weighing method butyrometers
    Stopper, rubber, for sealing the bulbs of all weighing method butyrometers
    614-2029
    Log in for availability
    1 * 100 ST - 24,10 €