ContactUsLinkComponent
Cream butyrometer, Schulz-Kley´s method
Cream butyrometer, Schulz-Kley´s method
Catalog # 614-0119
Supplier:  GERBER FUNKE
undefined
Cream butyrometer, Schulz-Kley´s method
Catalog # 614-0119
Supplier:  GERBER FUNKE
Specifications
  • Description:
    Butyrometer for cream, Schulz-Kley´s, 0 - 5 - 40 %: 0,5
  • Pk:
    10
Specifications
About this item
According to Schulz-Kley´s weighing method with closed bulb.