Orders LinkContactUsLinkComponent
Dissection set
Dissection set
Dissection set in a wooden case, 9 pcs.
Catalog # HERE1110401
Supplier:   HERENZ HEINZ
Dissection set
Catalog # HERE1110401
Supplier:  HERENZ HEINZ
Dissection set
Dissection set
Dissection set in a wooden case, 9 pcs.
Price
129,00 €
Log in for availability
1 * 1 ST
Product Details & Documents
This dissection set includes nine instruments in a strong wooden case.
Content:
1 scalpel handle No. 4 with two blades
1 scalpel
1 scalpel handle No. 3 with two blades
1 forceps
1 scissors (pointed)
2 dissection needles (1 pointed, 1 with blade)
Specifications
  • Description:
    Dissection set in a wooden case, 9 pcs.