ContactUsLinkComponent
Joining pieces for flat flanges, BOLA
Joining pieces for flat flanges, BOLA
Catalog # BOHLB323-01
Supplier:  Bohlender
undefined
Joining pieces for flat flanges, BOLA
Catalog # BOHLB323-01
Supplier:  Bohlender
Specifications
  • Description:
    Joining piece with 8 screws, silumin
  • Pk:
    1
Specifications
About this item
Made of aluminium.
  • For flanges NW 170/146
Connection between reaction vessel and lid. Locked by zinc-plated steel screws.