pH meters, handheld, PHT 810 / ST 1000
Fournisseur: EBRO
Certificats
À propos de cet article
This portable instrument measures the pH value in meat, cold cuts, cheese, dairy products and fluids. It is therefore ideal for butchers, super markets and in the beverage production.
- Handy and robust
- Simple calibration with keypad
- Replaceable electrode
- Current value memory
- Fully automatic pH-calibration
- Battery lifetime approx. 5 years
Livraison: The instrument PHT 810 is supplied without electrode. The set ST 1000 includes the pH meter PHT810, the penetration electrode AT 206, punching pin, buffer solution pH 4/7, protein cleaner, KCl-solution and case.
Spécifications
- Catalog No:
- EBRO1340-5810
- 620-2393
- Model:
- ST 1000
- PHT 810
- pH range:
- 0...+14,0
- 0...+14,0
- pH resolution:
- 0,01
- 0,01
- pH accuracy:
- ±0,03
- ±0,03
- Temperature range:
- −10,0...+50,0 °C
- −10,0...+50,0 °C
- Temperature compensation:
- Manual
- Manual
- Display:
- LCD, 12 mm
- LCD, 12 mm
- Memory:
- Hold, Min/Max
- Hold, Min/Max
- Connections:
- BNC
- BNC
- W×D×H:
- 110×54×22 mm
- 110×54×22 mm
- Weight:
- 0,2 kg
- 0,2 kg
Spécifications
Frequently Bought Together