Order Entry
Belgium
Orders LinkContactUsLinkComponent
Accessories

Accessories

Supplier:   Atago
Accessories
Supplier:  Atago
Certificates

11 Options Available Below

Product Details & Documents
Product Family Options

Product Information

  • Availability
    Price
  • ATAGRE-1600
    Prism, main and secondary for DR-M2
    Prism, main and secondary for DR-M2
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST

  • ATAGRE-1640
    Prism, main and secondary for DR-M4, DR-M4/1550
    Prism, main and secondary for DR-M4, DR-M4/1550
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST

  • ATAGRE-3521
    Interference filter, 486 nm (F), for DR-M2
    Interference filter, 486 nm (F), for DR-M2
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST

  • ATAGRE-3522
    Interference filter, 656 nm (C), for DR-M2
    Interference filter, 656 nm (C), for DR-M2
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST

  • ATAGRE-3523
    Interference filter, 546 nm (E), for DR-M2
    Interference filter, 546 nm (E), for DR-M2
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST

  • ATAGRE-3524
    Interference filter, 480 nm (F'), for DR-M2
    Interference filter, 480 nm (F'), for DR-M2
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST

  • ATAGRE-3525
    Interference filter, 644 nm (C'), for DR-M2
    Interference filter, 644 nm (C'), for DR-M2
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST

  • ATAGRE-3526/450
    Interference filter, 450 - 539 nm
    Interference filter, 450 - 539 nm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST

  • ATAGRE-3526/540
    Interference filter, 540 - 680 nm
    Interference filter, 540 - 680 nm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST

  • ATAGRE-3526/681
    Interference filter, 681 - 799 nm
    Interference filter, 681 - 799 nm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST

  • ATAGRE-3526/800
    Interference filter, 800 - 1100 nm
    Interference filter, 800 - 1100 nm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.