ContactUsLinkComponent
Evaporating dishes, PP, SP Bel-Art
Catalog # BELAF409250000
undefined
Evaporating dishes, PP, SP Bel-Art
Catalog # BELAF409250000
Specifications
  • Ext.Ø:
    100 mm
  • Capacity:
    175 ml
  • Height:
    42 mm
  • Pk:
    6
Specifications
About this item
Dishes, made of PP, translucent, smooth, with pour spout.
  • Useful for titration and evaporation
  • Resists acids and alkalis
  • Very good temperature resistance
Can also be used as a reusable weighing dish.