Liquid nitrogen level control, auto-fill system
À propos de cet article
Level control for liquid nitrogen type is manual or automatic refill. It is used for level control of liquid nitrogen (LN₂) in unsealed Dewar flasks.
- Parting of cable safety device for sensor
- Optical indication of the actual state
Applicable for acoustic and optical error indication, the controller opens whole refilling the magnetic valve at the reservoir tank.
Delivery information: Supplied with two temperature sensors PT 100 (minimum, maximum sensors), socket for LN₂-magnetic valve (24 V), function control for sensor.
2 Options disponibles ci-dessous
Product Details & Documents
Level control for liquid nitrogen type is manual or automatic refill. It is used for level control of liquid nitrogen (LN₂) in unsealed Dewar flasks.
- Parting of cable safety device for sensor
- Optical indication of the actual state
Applicable for acoustic and optical error indication, the controller opens whole refilling the magnetic valve at the reservoir tank.
Delivery information: Supplied with two temperature sensors PT 100 (minimum, maximum sensors), socket for LN₂-magnetic valve (24 V), function control for sensor.
Product Family Options
Informations sur le produit