Test tube racks, half-size, Poxygrid®, SP Bel-Art
Fournisseur: Bel-Art Products, a Part of SP
Certificats
À propos de cet article
Chemical and corrosion-resistant steel wire racks are coated with heat-cured epoxy.
- Unaffected by organic solvents, salts, weak acids and weak alkalis
- Will withstand a temperature range of −223 to 121 ºC; autoclavable
Excellent space-savers for use in water baths, laminar flow hoods, fume hoods or any place where space is limited.
Choice of three colours: orange for biohazard warning, green and blue for general lab use.
Frequently Bought Together