ContactUsLinkComponent
Rack for cuvettes, SP Bel-Art
Rack for cuvettes, SP Bel-Art
Catalog # BELAF185150000
undefined
Rack for cuvettes, SP Bel-Art
Catalog # BELAF185150000
Specifications
  • For:
    20 (2×10) cuvettes
  • W×D×H:
    210×70×35 mm
  • Pk:
    1
Specifications
About this item
Made of white PP. Rack holds 10 mm light path spectrophotometer cuvettes in two tiers with numbered wells.
  • Autoclavable
  • Very good chemical resiatnce
  • Easy to clean
The wells hold standard cuvettes with a 12,5 mm cross-section. A sloped top makes the second tier more accessible.