Order Entry
Belgium
ContactUsLinkComponent
Racks for centrifuge tubes, polystyrene friction-fit
Racks for centrifuge tubes, polystyrene friction-fit
Catalog # HEATHS23053A
Supplier:  HEATHROW
undefined
Racks for centrifuge tubes, polystyrene friction-fit
Catalog # HEATHS23053A
Supplier:  HEATHROW

Specifications

  • For tubes:
    50 ml
  • Hole Ø:
    27,5 - 29 mm
  • No. of Holes:
    1
  • Colour:
    Blue
  • Pk:
    5

Specifications

About this item

PS

  • Rack slides smoothly across work surfaces
  • Snug fit for tubes