Order Entry
Belgium
ContactUsLinkComponent
 
Reagent reservoirs, 12 channel, SP Bel-Art
  BELAF378801200
undefined
Reagent reservoirs, 12 channel, SP Bel-Art
  BELAF378801200

 

  • Packed:
    Individual
  • Sterile:
    +
  • Type:
    PP, with fitted lid, V-shaped
  • Capacity:
    12×5 ml
  • Colour:
    White
  • W×D×H:
    127×58×32 mm
  • Pk:
    8

 

 

PP. This chemical resistant 12 channel reagent reservoir can be used with single, 8 and 12 channel pipettes.

  • Wide profile provides added stability
  • Autoclavable at 121 °C
  • Individually sealed and sterilized (8 per bag, 6 bags per case)
  • V-shaped wells allow complete sample withdrawal
  • Each channel is numbered to facilitate serial dilution and holds 5 ml
  • Fitted lid included