Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Belgium
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Shields
Safety Shields
Accessories for SP Bel-Art safety splash shields
Accessories for SP Bel-Art safety splash shields
113-2501
:
Bel-Art Products, a Part of SP
Accessories for SP Bel-Art safety splash shields
113-2501
:
Bel-Art Products, a Part of SP
Description:
C clamp, opens to 63 mm, for mounting safety splash shields
Seq:
01
Pk:
1