Saisie de commande
Belgium
ContactUsLinkComponent
Caps for screw top vials, 12×32 mm, WHEATON®
Caps for screw top vials, 12×32 mm, WHEATON®
Catalog # WHEA242245
Supplier:  DWK Life Sciences
undefined
Caps for screw top vials, 12×32 mm, WHEATON®
Catalog # WHEA242245
Supplier:  DWK Life Sciences

Specifications

  • Type:
    Top seal cap
  • Désignation:
    Natural, 8-425, with 0,25 mm PP septa
  • Cdt:
    1.000

Specifications

About this item