Orders LinkContactUsLinkComponent
Pipette controller, RF3000®
Pipette controller, RF3000®
RF3000®, 1 - 100 ml
Catalog # HEATHS3000
Supplier:   HEATHROW
Pipette controller, RF3000®
Catalog # HEATHS3000
Supplier:  HEATHROW
Pipette controller, RF3000®
Pipette controller, RF3000®
RF3000®, 1 - 100 ml
Price
560,00 €
Log in for availability
1 * 1 ST
Product Details & Documents
This microprocessor controlled pipette controller can be used with glass and plastic pipettes from 1 to 100 ml.
  • Long-life li-ion battery, with quick charge-up to 20 hours continuous use
  • Universal grip and soft coated triggers with direction indicators, for right or left handed users
  • Ergonomically balanced in the hand with a loaded pipette
  • Bright LEDs indicate three operation modes: high, low, and gravity
  • Toggle operation by pressing the large mode button
  • Low battery indicator; can be recharged when in use
  • Five-year warranty unit
  • One-year warranty rechargeable battery
An adapter can be fitted to the nose cone to stabilise small volume and thinner pipettes.
Delivery information: Supplied with charging stand, wall-mountable/magnetic holder, nozzle adapter for use with small pipettes, Li-Ion rechargeable battery, three replacement 0,45 µm filters, five nose cones (blue, black, yellow, red, grey), and low voltage micro-USB connector (5VDC) and approx. 6 ft (183 cm) cable connected to hi-efficiency double-insulated AC wall outlet adapter 100 to 240 V, 50/60 Hz with four interchangeable plugs.
Specifications
  • Capacity:
    1 - 100 ml
  • Description:
    RF3000®
Accessories
Product Information
  • Catalog #
    Availability
    Price
  • Replacement Battery for RF3000
    Replacement Battery for RF3000
    HEATHS300502
    Log in for availability
    1 * 1 ST - 77,20 €
Product Information
  • Catalog #
    Availability
    Price
  • Replacement Power Adapter for RF3000
    Replacement Power Adapter for RF3000
    HEATHS300501
    Log in for availability
    1 * 1 ST - 151,00 €
Product Information
  • Catalog #
    Availability
    Price
  • Replacement Filters for RF3000
    Replacement Filters for RF3000
    HEATHS5000RFFIL
    Log in for availability
    1 * 5 ST - 63,20 €