Orders LinkContactUsLinkComponent
Test tube racks, Stack Rack™, SP Bel-Art
Test tube racks, Stack Rack™, SP Bel-Art
Test tube racks, Stack Rack™, SP Bel-Art
Certificates
Test tube racks, Stack Rack™, SP Bel-Art
Test tube racks, Stack Rack™, SP Bel-Art
2 Options Available Below
Product Details & Documents
PP, white
  • Sturdy and chemical resistant
  • Will not shatter or stain
  • With top shelf height of 70 mm
These rigid racks features interlocking end plates with handles for secure stacking when loaded with tubes up to 100 mm long. Rows are numbered and lettered for easy tube identification.
Product Family Options
Product Information
  • No. of Holes
    For tube Ø
    Material
    W×D×H
    Availability
    Price
  • 40 (4×10)
    13 - 16 mm
    PP
    270×112×133 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 104,00 €
    104,00 €
    1 * 1 ST
       104,00 €
  • 24 (3×8)
    25 - 30 mm
    PP
    297×127×133 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 109,00 €
    109,00 €
    1 * 1 ST
       109,00 €