ContactUsLinkComponent
Racks, two tier, for microcentrifuge tubes, SP Bel-Art
Racks, two tier, for microcentrifuge tubes, SP Bel-Art
Catalog # 211-2664
undefined
Racks, two tier, for microcentrifuge tubes, SP Bel-Art
Catalog # 211-2664
Specifications
  • For tubes:
    1,5 ml
  • No. of Holes:
    20 (2×10)
  • W×D×H:
    210×70×35 mm
  • Pk:
    1
Specifications
About this item
This two tier PP rack holds microcentrifuge tubes in numbered wells and tubes are held roughly protruding above the rack surface for easy handling.