ContactUsLinkComponent
Desiccator plates, high heat, Minerit HD, SP Bel-Art
Desiccator plates, high heat, Minerit HD, SP Bel-Art
Catalog # BELAF420380230
undefined
Desiccator plates, high heat, Minerit HD, SP Bel-Art
Catalog # BELAF420380230
Specifications
  • Ø:
    230 mm
  • Type:
    Fibre reinforced cement board
  • Seq:
    03
  • Pk:
    1
Specifications
About this item
Withstand temperatures up to 149 °C (300 °F). Contain no asbestos and are non combustible, stable and chemical resistant. Circular and perforated with 6 mm diameter holes on 25 mm centers in a straight line pattern.