Order Entry
Belgium
ContactUsLinkComponent
 
Desiccator cabinets, auto, horizontal, secador®, SP Bel-Art
  BELAF420740220
undefined
Desiccator cabinets, auto, horizontal, secador®, SP Bel-Art
  BELAF420740220

 

  • W×D×H ext.:
    518×414×341 mm
  • Description:
    Secador® Auto-Desiccator cabinet, clear, 54 l, model 4.0
  • Seq:
    01
  • Pk:
    1
  • Shelves:
    2
  • With:
    1 chamber featuring humidty control and air circulation, with hygrometer and vented adjustable shelves

 

 

Secador® 4.0 auto desiccator cabinets are made from moulded Durastar® co-polyester which blocks 99% of UV light and resists staining, crazing and chemical attack.

  • Large, pull down, oven style door with easy-to-view digital hygrometer
  • Two perforated shelves facilitate circulation of dry air
  • Colour-indicating silica gel allows quick visual confirmation of desiccant status

Protecting valuable humidity-sensitive items, such as reagents or electronics, is easy and fully automatic with Secador® Auto-Desiccator cabinets. Automatic dehumidification and desiccant regeneration cycle operates continuously every 20 minutes. Compared to other auto-desiccators that regenerate once every 4 to 6 hours, the Secador® Auto-Desiccators attains a low %rH level on start-up and consistently maintains low %rH, especially with frequent entry to the desiccator interior.

 : Supplied with power cord/transformer with input connector.