Orders LinkContactUsLinkComponent
Degassing Filters
Degassing Filters
Supplier:   ROBU GLASFILTER
Degassing Filters
Supplier:  ROBU GLASFILTER
Certificates
Degassing Filters
Degassing Filters
5 Options Available Below
Product Details & Documents
ROBU® degassing filter with integrated VitraPOR® filter ensures efficient solvent degassing with inert gases, combined with reliable filtration. Ideal for chromatography and analytical applications, it offers high chemical and thermal resistance.
  • VitraPOR® porous glass filter
  • Combined degassing and filtration function
  • Ideal for chromatography
  • Solvent degassing with inert gases
  • High chemical and thermal resistance
  • Efficient bubble-free operation
  • Autoclavable
ROBU degassing filters combine solvent degassing and fine filtration in one compact, efficient solution. Made from high-purity borosilicate glass with integrated VitraPOR filter, they ensure effective removal of dissolved gases while providing consistent particle retention.
Designed for use with inert gases such as helium or nitrogen, the filters enable bubble-free solvent delivery - essential for sensitive analytical applications like HPLC and chromatography. With excellent chemical resistance and thermal stability, ROBU degassing filters are reusable, autoclavable, and ideal for continuous operation in demanding laboratory environments.
Certifications: In compliance with ISO 4796-80 and ASTM E128-99 standards.
Product Family Options
Product Information
  • Material
    Porosity
    For tube I-Ø
    Ø
    Length
    Availability
    Price
  • ROBU18500
    Degassing filter
    Degassing filter
    Borosilicate glass 3.3
    0
    5 mm
    12 mm
    25/37 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 39,70 €
    39,70 €
    1 * 1 ST
       39,70 €
    Specifications:
    More
    Specifications
    Cat. No.
    ROBU18500
  • ROBU18501
    Degassing filter
    Degassing filter
    Borosilicate glass 3.3
    1
    5 mm
    12 mm
    25/37 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 39,70 €
    39,70 €
    1 * 1 ST
       39,70 €
    Specifications:
    More
    Specifications
    Cat. No.
    ROBU18501
  • ROBU18502
    Degassing filter
    Degassing filter
    Borosilicate glass 3.3
    2
    5 mm
    12 mm
    25/37 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 39,70 €
    39,70 €
    1 * 1 ST
       39,70 €
    Specifications:
    More
    Specifications
    Cat. No.
    ROBU18502
  • ROBU18503
    Degassing filter
    Degassing filter
    Borosilicate glass 3.3
    3
    5 mm
    12 mm
    25/37 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 39,70 €
    39,70 €
    1 * 1 ST
       39,70 €
    Specifications:
    More
    Specifications
    Cat. No.
    ROBU18503
  • ROBU18504
    Degassing filter
    Degassing filter
    Borosilicate glass 3.3
    4
    5 mm
    12 mm
    25/37 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 43,70 €
    43,70 €
    1 * 1 ST
       43,70 €
    Specifications:
    More
    Specifications
    Cat. No.
    ROBU18504
Adapter for degassing filter
Product Information
  • Catalog #
    Description
    Tubing int.Ø
    Availability
    Price
  • Adapter for tube, PTFE; Filter Ø: 5 mm
    ¹/₈" | 3 m/m
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 14,80 €
    14,80 €
    1 * 1 ST
       14,80 €