Orders LinkContactUsLinkComponent
Rods, filtering, BOLA
Supplier:   Bohlender
Rods, filtering, BOLA
Supplier:  Bohlender
Certificates
Rods, filtering, BOLA
Rods, filtering, BOLA
4 Options Available Below
Product Details & Documents
These filtering rods are made of microporous PTFE.
  • Resistant to chemicals
  • Temperature resistance from –200 to +250 °C
Diameter and length are nominal dimensions and can contain a machining tolerance.
Product Family Options
Product Information
  • Ø×H
    Pore size
    Availability
    Price
  • 28×100 mm
    5 μm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 113,00 €
    113,00 €
    1 * 1 ST
       113,00 €
  • 32×120 mm
    5 μm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 147,00 €
    147,00 €
    1 * 1 ST
       147,00 €
  • 28×100 mm
    10 μm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 142,00 €
    142,00 €
    1 * 1 ST
       142,00 €
  • 28×100 mm
    50 μm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 113,00 €
    113,00 €
    1 * 1 ST
       113,00 €