Order Entry
Belgium
ContactUsLinkComponent
 
Silicon standard solution, concentrate, 1000 mg Si in 14% sodium hydroxide solution (from SiCl₄) Titrisol® standard for AAS, Supelco®
  1.09947.0001
 :  Merck
 :  
Silicon standard solution, concentrate, 1000 mg Si in 14% sodium hydroxide solution (from SiCl₄) Titrisol® standard for AAS, Supelco®
  1.09947.0001
 :  Merck
 :  

 

  • Concentration:
    1000 mg
  • Matrix:
    14% NH₄OH
  • Pack type:
    Plastic ampoule
  • Pk:
    1 Ampoul
  • M-Clarity quality level:
    MQ100
  • Element:
    Silicon
  • Symbol:
    Si

 

 

 : Traceable to SRM from NIST