Manganese standard solution, concentrate, 1000 mg Mn in water (from MnCl₂) Titrisol® standard for AAS, Supelco®
1.09988.0001
: Merck
:
- Concentration:1000 mg
- Matrix:H₂O
- Pack type:Plastic ampoule
- Pk:1 Ampoul
- M-Clarity quality level:MQ100
- Element:Manganese
- Symbol:Mn
: Traceable to SRM from NIST