

Cacodylic acid sodium salt trihydrate
Supplier: Roth Carl
Certificates
About this item
CAS Number:6131-99-3
:124-65-2
Formula:(CH₃)₂As(O)ONa·3H₂O
MDL Number:MFCD00149079
:213.94 g/mol
Storage Temperature:Ambient
EINECS:204-708-2
UN:1688
ADR:6.1,II
:GHS08
:GHS07
Frequently Bought Together