Osmium standard solution, 1000 mg/L Os in 20% hydrochloric acid (from (NH₄)₂OsCl₆), standard for AAS, Specpure®
Catalog # 42239.AE
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Concentration:1000 mg/L
- Matrix:20% HCl
- Pack type:Glass bottle
- Pk:100 mL
- Environmentally Preferable:
- MDL No.:MFCD00011147
- Element:Osmium
- Symbol:Os
Specifications
About this item
Molecular Formula: (NH₄)₂OsCl₆ in 20% HCl
Physical Form: Liquid
UN No.: 1789
MDL No.: MFCD00011147
Certifications: Traceable to NIST