Cacodylic acid sodium salt, Millipore®
Catalog # 205541-100
Supplier: Merck Millipore (Calbiochem)
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:100 g
- Seq:3401
Specifications
About this item
:
:159.94 g/mol
EINECS:204-708-2
UN:1688
ADR:6.1,II
CAS Number:124-65-2
Formula:C₂H₆AsNaO₂