Order Entry
Belgium
ContactUsLinkComponent
Scandium standard solution, 1000 mg/L Sc in 7% nitric acid (from Sc₂O₃) Certipur® standard for ICP, Supelco®
Scandium standard solution, 1000 mg/L Sc in 7% nitric acid (from Sc₂O₃) Certipur® standard for ICP, Supelco®
Catalog # 1.70349.0100
Supplier:  Merck
CAS Number:  
undefined
Scandium standard solution, 1000 mg/L Sc in 7% nitric acid (from Sc₂O₃) Certipur® standard for ICP, Supelco®
Catalog # 1.70349.0100
Supplier:  Merck
CAS Number:  

Specifications

  • Concentration:
    1000 mg/L
  • Matrix:
    7% HNO₃
  • Pack type:
    Plastic bottle
  • Pk:
    100 mL
  • M-Clarity quality level:
    MQ100
  • Element:
    Scandium
  • Symbol:
    Sc

Specifications

About this item

Certifications: Traceable to SRM from NIST