Rhenium standard solution, 1000 mg/L Re in 5% nitric acid (from Re), standard for AAS, Specpure®
Molecular Formula: Re in 5% HNO₃
Physical Form: Liquid
UN No.: 3264
MDL No.: MFCD00011195
: Traceable to NIST
- Environmentally Preferable:
Frequently Bought Together