Order Entry
Belgium
ContactUsLinkComponent
Rhodium standard solution, 10 mg/L Rh in 0,5 M nitric acid (from Rh(NO₃)₃) Certipur® standard for ICP-MS, Supelco®
Rhodium standard solution, 10 mg/L Rh in 0,5 M nitric acid (from Rh(NO₃)₃) Certipur® standard for ICP-MS, Supelco®
Catalog # 1.08525.0100
Supplier:  Merck
CAS Number:  
undefined
Rhodium standard solution, 10 mg/L Rh in 0,5 M nitric acid (from Rh(NO₃)₃) Certipur® standard for ICP-MS, Supelco®
Catalog # 1.08525.0100
Supplier:  Merck
CAS Number:  

Specifications

  • Concentration:
    10 mg/L
  • Matrix:
    0,5 M HNO₃
  • Pack type:
    Plastic bottle
  • Pk:
    100 mL
  • M-Clarity quality level:
    MQ100
  • Element:
    Rhodium
  • Symbol:
    Rh

Specifications

About this item

Certifications: Traceable to SRM from NIST