Niobium standard solution, 1000 mg/L Nb in 2% hydrofluoric acid (from NbCl₅), standard for ICP, Specpure®
Supplier: Thermo Fisher Scientific
Certificates
About this item
Molecular Formula: NbCl₅ in 2% HF
Physical Form: Liquid
UN No.: 2922
MDL No.: MFCD00011126
Certifications: Traceable to NIST
Specifications
- MDL No.:MFCD00011126
Specifications
Related Information
Frequently Bought Together