Germanium standard solution, 1000 mg/L Ge in 5% nitric acid with hydrofluoric acid (max. 1%) (de Ge), standard for ICP, Specpure®
Molecular Formula: Ge in 5% HNO₃/tr. HF
Physical Form: Liquid
UN No.: 3264
MDL No.: MFCD00085310
Traceable to NIST
: Traceable to NIST
- Environmentally Preferable:
Frequently Bought Together