Germanium standard solution, 10000 mg/L Ge in 5% nitric acid with hydrofluoric acid (max. 1%) (de Ge), standard for ICP, Specpure®
Supplier: Thermo Fisher Scientific
Certificates
About this item
Molecular Formula: Ge in 5% HNO₃/tr. HF
Physical Form: Liquid
UN No.: 3264
MDL No.: MFCD00085310
Certifications: Traceable to NIST
Specifications
- Environmentally Preferable:
Specifications
Related Information
Frequently Bought Together