Germanium standard solution, 10 mg/L Ge in 5% nitric acid with hydrofluoric acid (max. 1%) (de Ge), standard for AAS, Specpure®
Catalog # 45254.AE
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Concentration:10 mg/L
- Matrix:5% HNO₃/tr HF
- Pack type:Glass bottle
- Pk:100 mL
- Environmentally Preferable:
- Element:Germanium
- Symbol:Ge
Specifications
About this item
Certifications: Traceable to NIST