Order Entry
Belgium
ContactUsLinkComponent
 
Herbicide reference standard, Desethyl atrazine 10 µg/ml in acetonitrile
Catalog # PROONE7051A
CAS Number:  
Herbicide reference standard, Desethyl atrazine 10 µg/ml in acetonitrile
Catalog # PROONE7051A
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    10 mL
  • CAS No.:
    6190-65-4
  • Description:
    Desethyl atrazine 10 µg/ml in acetonitrile
  • Formula:
    C₆H₁₀ClN₅
  • Description:
    Desethyl atrazine

Specifications

About this item