Order Entry
Belgium
ContactUsLinkComponent
 
Herbicide reference standard, Dinoseb (2-sec-Butyl-4,6-dinitrophenol) 500 µg/ml in methanol
  PROOSL20235
 :  
Herbicide reference standard, Dinoseb (2-sec-Butyl-4,6-dinitrophenol) 500 µg/ml in methanol
  PROOSL20235
 :  

 

  • Pack type:
    Glass bottle
  • Pk:
    1 mL
  • CAS No.:
    88-85-7
  • Description:
    Dinoseb (2-sec-Butyl-4,6-dinitrophenol) 500 µg/ml in methanol
  • Formula:
    C₁₀H₁₂N₂O₅
  • Description:
    Dinoseb (2-sec-Butyl-4,6-dinitrophenol)