Selenium standard solution, 1000 mg/L Se in 5% nitric acid (from Se), standard for ICP, Specpure®
Supplier: Thermo Fisher Scientific
Certificates
About this item
Molecular Formula: Se in 5% HNO₃
Physical Form: Liquid
UN No.: 3264
MDL No.: MFCD00134090
Certifications: Traceable to NIST
Specifications
- Environmentally Preferable:
Specifications
Related Information
Frequently Bought Together