Scandium standard solution, 1000 mg/L Sc in 1 M nitric acid (from Sc₂O₃) Certipur® standard for AAS, Supelco®
: Merck
Scandium standard solution, 1000 mg/L Sc in 1 M nitric acid (from Sc₂O₃) Certipur® standard for AAS, Supelco®
: Merck
: Traceable to SRM from NIST
- M-Clarity quality level:MQ100
Frequently Bought Together
VWR® AVS TITRINORM Buffer Solutions, 20 °C
: VWR Chemicals
Membrane filters, cellulose nitrate, non sterile, Whatman™
: Whatman products (Cytiva)
Acetone ≥99%, TECHNICAL
: VWR Chemicals
VWR®, Nitrile Gloves
: VWR Collection